Learn More
Waters Corp Deactivated Clear Glass 12 x 32 mm Screw Neck Vial with Cap and Preslit PTFE/Silicone Septum, 2 mL Volume, 100/pk
Catalog No. 50818720 Shop All Waters Corporation Products
Encompass_Preferred
Deactivated vials have a silanized surface to prevent adsorption of highly polar molecules.
Catalog No. 50-818-720 Supplier Waters Corp Supplier No. 186000307DV
Deactivated vials have a silanized surface to prevent adsorption of highly polar molecules.