Learn More
Waters Corp Waters Vial, Deactivated Clear Glass, Screw Neck, 12 x 32 mm, 2 mL Volume
Catalog No. 50782504 Shop All Waters Corporation Products
Encompass_Preferred
This Deactivated Clear Glass 12 x 32 mm Screw Neck Vial is designed for safely storing polar analytes including phenols, proteins, and pesticides. It features a silanized surface to prevent adsorption of highly polar molecules and is treated with gas-phase reactive organosilane to produce a hydrophobic glass surface, ensuring chemical inertness.
  • Silanized surface prevents adsorption of highly polar molecules.
  • Specially treated to eliminate absorption of compounds onto the glass surface.
  • Produces a hydrophobic glass surface with gas-phase reactive organosilane treatment.
  • Compatible with glass-sensitive compounds and chemically inert.
  • Permanent surface modification ensures indefinite shelf life.
  • Reduces effects of extractables and prevents leaching.
  • Manufactured to ISO 9001, ISO 13485, & current Good Manufacturing Practices (CGMP) standards.
Catalog No. 50-782-504 Supplier Waters Corp Supplier No. 186000272DV
This Deactivated Clear Glass 12 x 32 mm Screw Neck Vial is designed for safely storing polar analytes including phenols, proteins, and pesticides. It features a silanized surface to prevent adsorption of highly polar molecules and is treated with gas-phase reactive organosilane to produce a hydrophobic glass surface, ensuring chemical inertness.
  • Silanized surface prevents adsorption of highly polar molecules.
  • Specially treated to eliminate absorption of compounds onto the glass surface.
  • Produces a hydrophobic glass surface with gas-phase reactive organosilane treatment.
  • Compatible with glass-sensitive compounds and chemically inert.
  • Permanent surface modification ensures indefinite shelf life.
  • Reduces effects of extractables and prevents leaching.
  • Manufactured to ISO 9001, ISO 13485, & current Good Manufacturing Practices (CGMP) standards.