Learn More
Waters Corp Waters Corporation Autosampler Vial, Deactivated Amber Glass, 12 x 32 mm, 2 mL
Catalog No. 50782529 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This deactivated amber glass 12 x 32 mm screw neck vial prevents sensitive samples from deteriorating. It offers compositional stability, a tight structure for analytical equipment, and chemical inertness. Each batch is UV-tested by HPLC to detect trace chemicals and ensure cleanliness. Includes a universal screw cap, with amber glass protecting against UV light.
  • Prevents sensitive samples from deteriorating.
  • Tight structure for secure fit with analytical equipment.
  • Stable to withstand chemical and mechanical forces.
  • Chemically inert, ensuring no interference.
  • UV-tested by HPLC to detect trace chemicals.
  • Each batch tested for cleanliness.
  • Equipped with a universal screw cap.
  • Amber color protects against UV light.
Catalog No. 50-782-529 Supplier Waters Corp Supplier No. 186000848DV
This deactivated amber glass 12 x 32 mm screw neck vial prevents sensitive samples from deteriorating. It offers compositional stability, a tight structure for analytical equipment, and chemical inertness. Each batch is UV-tested by HPLC to detect trace chemicals and ensure cleanliness. Includes a universal screw cap, with amber glass protecting against UV light.
  • Prevents sensitive samples from deteriorating.
  • Tight structure for secure fit with analytical equipment.
  • Stable to withstand chemical and mechanical forces.
  • Chemically inert, ensuring no interference.
  • UV-tested by HPLC to detect trace chemicals.
  • Each batch tested for cleanliness.
  • Equipped with a universal screw cap.
  • Amber color protects against UV light.