Learn More
Waters Corp Deactivated Amber Glass Snap Neck Vial, 12 x 32mm, 2 mL
Catalog No. 50785986 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This product is a deactivated amber glass vial designed for laboratory use. It has dimensions of 12 x 32 mm and a 2 mL volume. These vials are compatible with a wide range of analytical instruments from various manufacturers, ensuring broad applicability in laboratory settings.
  • Deactivated amber glass material.
  • 12 mm diameter.
  • 32 mm height.
  • 2 mL volume.
  • Snap neck vial type.
  • Supplied in a pack of 100.
  • Compatible with Agilent, Beckman, Dynatech, Finnigan, Fisons, Gilson, Hitachi, LDC, Perkin-Elmer, Spectra-Physics, Varian, Waters 2695, and Waters 2795 instruments.
Catalog No. 50-785-986 Supplier Waters Corp Supplier No. WAT094220DV
This product is a deactivated amber glass vial designed for laboratory use. It has dimensions of 12 x 32 mm and a 2 mL volume. These vials are compatible with a wide range of analytical instruments from various manufacturers, ensuring broad applicability in laboratory settings.
  • Deactivated amber glass material.
  • 12 mm diameter.
  • 32 mm height.
  • 2 mL volume.
  • Snap neck vial type.
  • Supplied in a pack of 100.
  • Compatible with Agilent, Beckman, Dynatech, Finnigan, Fisons, Gilson, Hitachi, LDC, Perkin-Elmer, Spectra-Physics, Varian, Waters 2695, and Waters 2795 instruments.