Learn More
Waters Corp Waters Screw Neck Vial, Deactivated Amber Glass, 12 x 32 mm, 2 mL Volume
Catalog No. 50782526 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This deactivated amber glass vial safely holds sensitive analytes, samples, and solutions. Its amber color prevents deterioration, and its compositional stability withstands chemical and mechanical stresses. The chemically tested and approved deactivated glass surface, treated with gas-phase reactive organosilane, is ideal for extremely polar analytes.
  • Deactivated amber glass construction prevents deterioration and is ideal for extremely polar analytes.
  • Chemically inert with compositional stability, enduring chemical and mechanical stresses.
  • Manufactured to ISO 9001 standards, with autosampler-specific dimensional tolerances confirmed.
  • Includes PTFE septum suitable for single injection applications and MS due to high solvent resistance.
  • Universal screw cap provides an excellent seal.
Catalog No. 50-782-526 Supplier Waters Corp Supplier No. 186000846DV
This deactivated amber glass vial safely holds sensitive analytes, samples, and solutions. Its amber color prevents deterioration, and its compositional stability withstands chemical and mechanical stresses. The chemically tested and approved deactivated glass surface, treated with gas-phase reactive organosilane, is ideal for extremely polar analytes.
  • Deactivated amber glass construction prevents deterioration and is ideal for extremely polar analytes.
  • Chemically inert with compositional stability, enduring chemical and mechanical stresses.
  • Manufactured to ISO 9001 standards, with autosampler-specific dimensional tolerances confirmed.
  • Includes PTFE septum suitable for single injection applications and MS due to high solvent resistance.
  • Universal screw cap provides an excellent seal.