Learn More
Waters Corp Waters Corporation Autosampler Vial, Deactivated Clear Glass, 12 x 32 mm, Screw Neck, 1 mL
Catalog No. 50782517 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This deactivated clear glass screw neck vial safely stores polar analytes (e.g., phenols, proteins). Its gas-phase reactive organosilane deactivation creates a permanent, hydrophobic glass surface, ensuring indefinite shelf life and eliminating sample interaction. Manufactured to ISO 9001, ISO 13485, and cGMP standards, it's compatible with many autosamplers and offers air-tight, leak-free seals.
  • Deactivated clear glass prevents polar analyte adsorption
  • Hydrophobic surface from organosilane treatment
  • Permanent surface modification for indefinite shelf life
  • Ideal for sensitive samples (e.g., biological, pharmaceutical)
  • Manufactured to ISO 9001, ISO 13485, and cGMP standards
  • Compatible with wide range of autosamplers
  • Includes cap and preslit PTFE/Silicone septum
Catalog No. 50-782-517 Supplier Waters Corp Supplier No. 186000385DV
This deactivated clear glass screw neck vial safely stores polar analytes (e.g., phenols, proteins). Its gas-phase reactive organosilane deactivation creates a permanent, hydrophobic glass surface, ensuring indefinite shelf life and eliminating sample interaction. Manufactured to ISO 9001, ISO 13485, and cGMP standards, it's compatible with many autosamplers and offers air-tight, leak-free seals.
  • Deactivated clear glass prevents polar analyte adsorption
  • Hydrophobic surface from organosilane treatment
  • Permanent surface modification for indefinite shelf life
  • Ideal for sensitive samples (e.g., biological, pharmaceutical)
  • Manufactured to ISO 9001, ISO 13485, and cGMP standards
  • Compatible with wide range of autosamplers
  • Includes cap and preslit PTFE/Silicone septum