Learn More
Waters Corp WATERS CORPORATION Screw Neck Vial, Deactivated Clear Glass, 12 x 32 mm, 2 mL Volume
Catalog No. 50782505 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Deactivated vials have a silanized surface to prevent adsorption of highly polar molecules. These high-quality glass vials are essential for a range of applications, ensuring uncompromised sample analysis. Treated with a reactive organosilane, the hydrophobic glass surface prevents compound adsorption, maintaining the integrity of your samples. Featuring a convenient screw neck closure, these vials are effortless to open and close, guaranteeing ease of use in your lab equipment.
  • Crafted from deactivated clear glass, exhibiting exceptional chemical resistance and durability.
  • Hydrophobic glass surface effectively prevents the adsorption of compounds, eliminating interference with accurate analysis.
  • Screw neck closure offers effortless access to the vials, streamlining laboratory workflows.
  • LC and GC certification guarantees compatibility with these analytical methods.
Catalog No. 50-782-505 Supplier Waters Corp Supplier No. 186000273DV
Deactivated vials have a silanized surface to prevent adsorption of highly polar molecules. These high-quality glass vials are essential for a range of applications, ensuring uncompromised sample analysis. Treated with a reactive organosilane, the hydrophobic glass surface prevents compound adsorption, maintaining the integrity of your samples. Featuring a convenient screw neck closure, these vials are effortless to open and close, guaranteeing ease of use in your lab equipment.
  • Crafted from deactivated clear glass, exhibiting exceptional chemical resistance and durability.
  • Hydrophobic glass surface effectively prevents the adsorption of compounds, eliminating interference with accurate analysis.
  • Screw neck closure offers effortless access to the vials, streamlining laboratory workflows.
  • LC and GC certification guarantees compatibility with these analytical methods.