Learn More
Waters Corp Waters Insert, Deactivated Clear Glass, 150 μL Volume, with Polyspring
Catalog No. 50785980 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Deactivated Clear Glass Insert is a critical LC accessory designed to prevent the adsorption of highly polar molecules by utilizing a silanized surface. This silanization process deactivates the glass, eliminating its absorptive character and ensuring high-quality analysis and safe storage of polar molecules.
  • Silanized surface to prevent adsorption of highly polar molecules.
  • Ideal for highly polar analytes that may associate with standard polar glass surfaces.
  • Bottom thickness held to an extremely narrow tolerance to eliminate needling damage.
  • Treated with gas-phase, reactive organosilane, producing a hydrophobic glass surface.
  • Can be stored dry indefinitely, reducing storage maintenance costs.
Catalog No. 50-785-980 Supplier Waters Corp Supplier No. WAT094171DV
The Deactivated Clear Glass Insert is a critical LC accessory designed to prevent the adsorption of highly polar molecules by utilizing a silanized surface. This silanization process deactivates the glass, eliminating its absorptive character and ensuring high-quality analysis and safe storage of polar molecules.
  • Silanized surface to prevent adsorption of highly polar molecules.
  • Ideal for highly polar analytes that may associate with standard polar glass surfaces.
  • Bottom thickness held to an extremely narrow tolerance to eliminate needling damage.
  • Treated with gas-phase, reactive organosilane, producing a hydrophobic glass surface.
  • Can be stored dry indefinitely, reducing storage maintenance costs.