Learn More
Waters Corp Waters Replacement Inserts, Deactivated Glass, 96-well Plate, 1 mL
Catalog No. 50782549 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These quick load, 1 mL deactivated glass inserts are designed for use with 96-well plates, offering a solution for applications with limited sample volume. The deactivated glass features a silanized surface to prevent the adsorption of highly polar molecules, and its inert construction helps maintain analytical result integrity.
  • Quick load design for convenient use.
  • Deactivated glass insert with silanized surface prevents adsorption of highly polar molecules.
  • Ideal for applications with limited sample volume.
  • Manufactured using ultra-pure reagents in an ISO-certified facility.
  • Batch examined for outstanding, repeatable performance.
  • Constructed of inert glass to maintain analytical results integrity.
  • Suitable for HPLC platforms in pharmaceuticals, environmental, forensics, and biopharmaceutical applications.
  • Provided as a single unit per package.
Catalog No. 50-782-549 Supplier Waters Corp Supplier No. 186001436DV
These quick load, 1 mL deactivated glass inserts are designed for use with 96-well plates, offering a solution for applications with limited sample volume. The deactivated glass features a silanized surface to prevent the adsorption of highly polar molecules, and its inert construction helps maintain analytical result integrity.
  • Quick load design for convenient use.
  • Deactivated glass insert with silanized surface prevents adsorption of highly polar molecules.
  • Ideal for applications with limited sample volume.
  • Manufactured using ultra-pure reagents in an ISO-certified facility.
  • Batch examined for outstanding, repeatable performance.
  • Constructed of inert glass to maintain analytical results integrity.
  • Suitable for HPLC platforms in pharmaceuticals, environmental, forensics, and biopharmaceutical applications.
  • Provided as a single unit per package.