Learn More
Waters Corp Waters Insert, Deactivated, Clear Glass, 250 μL Volume
Catalog No. 50785073 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Deactivated vials have a silanized surface to prevent adsorption of highly polar molecules.
  • Deactivated surface prevents adsorption of highly polar molecules.
  • Constructed from clear glass.
  • Features a 250 μL volume.
  • Suitable for sample preparation.
  • Supplied in packages of 144 units.
Catalog No. 50-785-073 Supplier Waters Corp Supplier No. WAT015199DV
Deactivated vials have a silanized surface to prevent adsorption of highly polar molecules.
  • Deactivated surface prevents adsorption of highly polar molecules.
  • Constructed from clear glass.
  • Features a 250 μL volume.
  • Suitable for sample preparation.
  • Supplied in packages of 144 units.