Learn More
Waters Corp Waters Corporation Insert, Deactivated, Clear Glass, 250 μL Volume
Catalog No. 50785888 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These deactivated clear glass inserts are designed for sample preparation applications. They feature a 250 μL volume and are provided in a package of 100.
  • Includes insert
  • Suitable for sample limited applications
  • Provided in packages of 100 units
Catalog No. 50-785-888 Supplier Waters Corp Supplier No. WAT072704DV
These deactivated clear glass inserts are designed for sample preparation applications. They feature a 250 μL volume and are provided in a package of 100.
  • Includes insert
  • Suitable for sample limited applications
  • Provided in packages of 100 units