Learn More
Waters Corp Waters Glass Insert, Deactivated, Clear, 300 μL Volume, with Polyspring
Catalog No. 50785978 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
When working with limited sample volumes, vial inserts are an effective choice to help adjust other lab equipment such as sample plates. By reducing the volume, the surface area is also lowered, offering ideal conditions for the needle when withdrawing samples. The deactivated glass used for this vial has a silanized surface designed to prevent the adsorption of highly polar molecules.
  • Deactivated glass prevents adsorption of highly polar molecules, proteins, and peptides.
  • Silanized surface is treated with reactive organosilane, creating a hydrophobic glass surface.
  • Reduces sample volume and surface area, providing ideal conditions for needle withdrawal.
  • Includes a polyspring.
  • Can be stored dry indefinitely.
  • Constructed from Type 1, 33-Expansions borosilicate glass for chemical inertness and high-quality results.
  • Compatible with many proprietary vials.
Catalog No. 50-785-978 Supplier Waters Corp Supplier No. WAT094170DV
When working with limited sample volumes, vial inserts are an effective choice to help adjust other lab equipment such as sample plates. By reducing the volume, the surface area is also lowered, offering ideal conditions for the needle when withdrawing samples. The deactivated glass used for this vial has a silanized surface designed to prevent the adsorption of highly polar molecules.
  • Deactivated glass prevents adsorption of highly polar molecules, proteins, and peptides.
  • Silanized surface is treated with reactive organosilane, creating a hydrophobic glass surface.
  • Reduces sample volume and surface area, providing ideal conditions for needle withdrawal.
  • Includes a polyspring.
  • Can be stored dry indefinitely.
  • Constructed from Type 1, 33-Expansions borosilicate glass for chemical inertness and high-quality results.
  • Compatible with many proprietary vials.