Learn More
Waters Corp Deactivated Amber Glass 15 x 45 mm Screw Neck Vial, with Cap and Preslit PTFE/Silicone Septum, 4 mL Volume, 100/pk
Catalog No. 50782547 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Deactivated vials have a silanized surface to prevent adsorption of highly polar molecules.
Catalog No. 50-782-547 Supplier Waters Corp Supplier No. 186001134DV
Deactivated vials have a silanized surface to prevent adsorption of highly polar molecules.