Learn More
Waters Corp Waters Vial, Deactivated Clear Glass, 15 x 45 mm, 4 mL
Catalog No. 50782524 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These deactivated clear glass vials are designed to prevent adsorption of highly polar molecules, making them ideal for storing analytes such as phenols, proteins, antibodies, amines, metabolites, herbicides, and pesticides. A gas-phase reactive organosilane treatment creates a permanent, hydrophobic, and chemically inert surface, preventing compound absorption and minimizing extractables. This reduces sample interaction and ensures sample integrity.
  • Silanized surface prevents adsorption of highly polar molecules.
  • Hydrophobic & chemically inert glass surface from organosilane treatment.
  • Permanent surface modification ensures consistent performance.
  • Minimizes extractables & leakage into sample.
  • Reduces sample interaction; ideal for biological, pharmaceutical, natural product, pesticide, & herbicide analyses.
  • Manufactured to ISO 9001, ISO 13485, & CGMP standards.
Catalog No. 50-782-524 Supplier Waters Corp Supplier No. 186000840DV
These deactivated clear glass vials are designed to prevent adsorption of highly polar molecules, making them ideal for storing analytes such as phenols, proteins, antibodies, amines, metabolites, herbicides, and pesticides. A gas-phase reactive organosilane treatment creates a permanent, hydrophobic, and chemically inert surface, preventing compound absorption and minimizing extractables. This reduces sample interaction and ensures sample integrity.
  • Silanized surface prevents adsorption of highly polar molecules.
  • Hydrophobic & chemically inert glass surface from organosilane treatment.
  • Permanent surface modification ensures consistent performance.
  • Minimizes extractables & leakage into sample.
  • Reduces sample interaction; ideal for biological, pharmaceutical, natural product, pesticide, & herbicide analyses.
  • Manufactured to ISO 9001, ISO 13485, & CGMP standards.