Learn More
Waters Corp Waters Corporation Autosampler Vial, Deactivated Clear Glass, Screw Neck, 15 x 45 mm, 4 mL
Catalog No. 50782523 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Deactivated vials have a silanized surface to prevent adsorption of highly polar molecules.
  • Deactivated surface to prevent adsorption of highly polar molecules.
  • Includes vial, cap, and septum.
  • Screw neck closure for secure sealing.
  • Dimensions of 15 x 45 mm.
  • 4 mL volume capacity.
  • Suitable for autosampler applications.
  • Ideal for sample preparation.
Catalog No. 50-782-523 Supplier Waters Corp Supplier No. 186000839DV
Deactivated vials have a silanized surface to prevent adsorption of highly polar molecules.
  • Deactivated surface to prevent adsorption of highly polar molecules.
  • Includes vial, cap, and septum.
  • Screw neck closure for secure sealing.
  • Dimensions of 15 x 45 mm.
  • 4 mL volume capacity.
  • Suitable for autosampler applications.
  • Ideal for sample preparation.