Learn More
Waters Corp Waters Corporation Replacement Inserts, Deactivated, 700 μL, Glass, 96-well Plate
Catalog No. 50782550 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These replacement inserts are a laboratory essential designed for applications with limited sample volume. Each 700 μL deactivated glass insert is for use with 96-well plates. The deactivated glass features a silanized surface, crucial for preventing the adsorption of highly polar molecules and ensuring reliable sample integrity. They are produced in facilities adhering to ISO 9001, ISO 13485, and cGMP standards, with rigorous testing and quality control for consistent results.
  • Quick load design.
  • 700 μL deactivated glass insert.
  • Silanized surface prevents adsorption of highly polar molecules.
  • Manufactured in dedicated facilities meeting stringent industry standards (ISO 9001, ISO 13485, cGMP).
  • Rigorously tested and passes quality control for consistent results.
Catalog No. 50-782-550 Supplier Waters Corp Supplier No. 186001437DV
Add to Cart
Add to Cart
These replacement inserts are a laboratory essential designed for applications with limited sample volume. Each 700 μL deactivated glass insert is for use with 96-well plates. The deactivated glass features a silanized surface, crucial for preventing the adsorption of highly polar molecules and ensuring reliable sample integrity. They are produced in facilities adhering to ISO 9001, ISO 13485, and cGMP standards, with rigorous testing and quality control for consistent results.
  • Quick load design.
  • 700 μL deactivated glass insert.
  • Silanized surface prevents adsorption of highly polar molecules.
  • Manufactured in dedicated facilities meeting stringent industry standards (ISO 9001, ISO 13485, cGMP).
  • Rigorously tested and passes quality control for consistent results.