Learn More
Waters Corp Deactivated Clear Glass 8 x 40 mm Snap Neck Vial, with Cap and PTFE Septum, 1 mL Volume, 250/pk
Catalog No. 50785208 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Deactivated vials have a silanized surface to prevent adsorption of highly polar molecules.
Catalog No. 50-785-208 Supplier Waters Corp Supplier No. WAT025054DV
Deactivated vials have a silanized surface to prevent adsorption of highly polar molecules.