Learn More
Waters Corp Waters Crimp Vial, Deactivated Amber Glass, 12 x 32 mm, 2 mL
Catalog No. 50785990 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This vial features a silanized surface to prevent adsorption of highly polar molecules, and is specifically designed to protect sensitive samples from light and UV radiation. Versatile in its application, it can accommodate both liquid and solid samples and is compatible with most common autosamplers.
  • Designed to protect sensitive samples from light and UV radiation.
  • Versatile for accommodating both liquid and solid samples.
  • Supplied with a crimp cap, ensuring a secure seal.
  • Compatible with most common autosamplers.
  • Offers a robust and reliable sealing solution for enhanced protection and integrity of samples.
Catalog No. 50-785-990 Supplier Waters Corp Supplier No. WAT094223DV
This vial features a silanized surface to prevent adsorption of highly polar molecules, and is specifically designed to protect sensitive samples from light and UV radiation. Versatile in its application, it can accommodate both liquid and solid samples and is compatible with most common autosamplers.
  • Designed to protect sensitive samples from light and UV radiation.
  • Versatile for accommodating both liquid and solid samples.
  • Supplied with a crimp cap, ensuring a secure seal.
  • Compatible with most common autosamplers.
  • Offers a robust and reliable sealing solution for enhanced protection and integrity of samples.