Learn More
Waters Corp Waters Vial, Deactivated Amber Glass, Screw Neck, 300 μL, Qsert
Catalog No. 50782541 Shop All Waters Corporation Products
Encompass_Preferred
This 12 x 32 mm screw neck vial is specifically designed for limited sample volumes. It features a glass insert within the vial that measures 300 μL. Each vial is strictly tested for cleanliness and dimensional tolerances, particularly for the Qsert insert, to ensure optimal sample volume and purity. The deactivated amber glass provides unique UV-blocking properties to protect samples susceptible to UV degradation during sample preparation assays, which is crucial when working with limited sample volumes where degradation cannot be afforded.
  • Unique UV-blocking properties to protect samples from UV degradation.
  • Designed for limited sample volumes with a 300 μL glass insert.
  • Strictly tested for cleanliness and dimensional tolerances.
  • Comes with a cap and PTFE/silicone septum.
  • Autosampler vial type.
  • Suitable for sample preparation applications.
  • 100 units per package.
Catalog No. 50-782-541 Supplier Waters Corp Supplier No. 186001130DV
This 12 x 32 mm screw neck vial is specifically designed for limited sample volumes. It features a glass insert within the vial that measures 300 μL. Each vial is strictly tested for cleanliness and dimensional tolerances, particularly for the Qsert insert, to ensure optimal sample volume and purity. The deactivated amber glass provides unique UV-blocking properties to protect samples susceptible to UV degradation during sample preparation assays, which is crucial when working with limited sample volumes where degradation cannot be afforded.
  • Unique UV-blocking properties to protect samples from UV degradation.
  • Designed for limited sample volumes with a 300 μL glass insert.
  • Strictly tested for cleanliness and dimensional tolerances.
  • Comes with a cap and PTFE/silicone septum.
  • Autosampler vial type.
  • Suitable for sample preparation applications.
  • 100 units per package.