Learn More
Waters Corp Deactivated Glass Insert, 200 μL Volume, Borosilicate Glass
Catalog No. 502390666 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Deactivated Glass Insert is a reusable tool designed for sample preparation applications. It has a 200 μL capacity and is made from high-quality borosilicate glass, offering resistance to breakage and contamination. The insert undergoes a deactivation process through heat treatment to remove active sites that could adsorb analytes, aiming for accurate and reliable results.
  • Volume capacity of 200 μL
  • Crafted from high-quality borosilicate glass
  • Deactivation process minimizes analyte adsorption
  • Designed for easy cleaning and multiple uses
  • Exceptional resistance to breakage and contamination
  • Fits most standard 2 mL autosampler vials
  • Minimizes sample loss and enhances reproducibility in chromatographic runs
  • Suitable for liquid chromatography, gas chromatography, and mass spectrometry
Catalog No. 50-239-0666 Supplier Waters Corp Supplier No. WAT072294DV
The Deactivated Glass Insert is a reusable tool designed for sample preparation applications. It has a 200 μL capacity and is made from high-quality borosilicate glass, offering resistance to breakage and contamination. The insert undergoes a deactivation process through heat treatment to remove active sites that could adsorb analytes, aiming for accurate and reliable results.
  • Volume capacity of 200 μL
  • Crafted from high-quality borosilicate glass
  • Deactivation process minimizes analyte adsorption
  • Designed for easy cleaning and multiple uses
  • Exceptional resistance to breakage and contamination
  • Fits most standard 2 mL autosampler vials
  • Minimizes sample loss and enhances reproducibility in chromatographic runs
  • Suitable for liquid chromatography, gas chromatography, and mass spectrometry