Learn More
Waters Corp Waters Corporation Screw Neck Vial, Deactivated Clear Glass, 12 x 32 mm, 2 mL
Catalog No. 50782532 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These deactivated clear glass screw neck vials prevent adsorption of highly polar molecules, ensuring sample integrity. With a pre-slit PTFE/silicone septum and screw cap, they provide a secure, leak-resistant seal. Pre-assembled for dissolution systems, they are ideal for multiple injections and long-term storage.
  • Silanized surface prevents highly polar molecule adsorption.
  • Screw neck design with pre-slit PTFE/silicone septum for secure seal.
  • Aids analyte transfer, reduces sample loss.
  • Pre-assembled for dissolution systems.
  • PTFE/silicone septum offers excellent resealing.
  • Septum operates –40°C to 200°C.
  • Manufactured with inert materials for interference-free results.
  • Caps protect against leakage, ensure long shelf life.
Catalog No. 50-782-532 Supplier Waters Corp Supplier No. 186000989DV
These deactivated clear glass screw neck vials prevent adsorption of highly polar molecules, ensuring sample integrity. With a pre-slit PTFE/silicone septum and screw cap, they provide a secure, leak-resistant seal. Pre-assembled for dissolution systems, they are ideal for multiple injections and long-term storage.
  • Silanized surface prevents highly polar molecule adsorption.
  • Screw neck design with pre-slit PTFE/silicone septum for secure seal.
  • Aids analyte transfer, reduces sample loss.
  • Pre-assembled for dissolution systems.
  • PTFE/silicone septum offers excellent resealing.
  • Septum operates –40°C to 200°C.
  • Manufactured with inert materials for interference-free results.
  • Caps protect against leakage, ensure long shelf life.