Learn More
Waters Corp Qsert Vial, Deactivated Clear Glass, 12 x 32 mm, Snap Neck
Catalog No. 50782537 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Qsert Vial features deactivated clear glass, measuring 12 x 32 mm, and has a snap neck design. It is supplied with a cap and PTFE septum, offering a 300 μL working volume. These vials are suitable for sensitive analytical applications where adsorption of polar molecules needs to be minimized.
  • Deactivated clear glass construction.
  • Measures 12 x 32 mm.
  • Snap neck design.
  • Includes cap and PTFE septum.
  • Working volume of 300 μL.
  • Supplied in a pack of 100.
Catalog No. 50-782-537 Supplier Waters Corp Supplier No. 186001127DV
This Qsert Vial features deactivated clear glass, measuring 12 x 32 mm, and has a snap neck design. It is supplied with a cap and PTFE septum, offering a 300 μL working volume. These vials are suitable for sensitive analytical applications where adsorption of polar molecules needs to be minimized.
  • Deactivated clear glass construction.
  • Measures 12 x 32 mm.
  • Snap neck design.
  • Includes cap and PTFE septum.
  • Working volume of 300 μL.
  • Supplied in a pack of 100.