Learn More
Waters Corp Waters Vial, Deactivated Clear Glass, 12 x 32 mm, Snap Neck, 300 μL Volume
Supplier:  Waters Corp 186001125DV
Encompass_Preferred
This deactivated clear glass snap neck Qsert vial is designed for autosamplers and sensitive samples. It features a 12 x 32 mm dimension and a 300 μL volume, including a cap and a preslit PTFE/silicone septum.
  • Deactivated clear glass provides an inert surface for sensitive samples and clear visibility of contents.
  • Snap neck facilitates easy and secure capping.
  • Qsert vial format is optimized for specific sample handling or autosampler compatibility.
  • Preslit PTFE/silicone septum offers convenient and reliable sealing, good chemical resistance, and ease of needle penetration for autosamplers.
  • Designed for use in automated sample injection systems, ensuring compatibility and efficient workflow.
Catalog No. 50-782-534