Learn More
Waters Corp Waters Vial, Deactivated Clear Glass, Max Recovery, Screw Neck, 12 x 32 mm, 1.5 mL
Supplier:  Waters Corp 186000326DV
Encompass_Preferred
Deactivated vials have a silanized surface to prevent adsorption of highly polar molecules. The Deactivated Clear Glass Screw Neck Max Recovery Vial measures 12 x 32 mm and offers a cap and PTFE/silicone septum. It is designed to withhold 1.5 mL in sample volume and provides the opportunity for clean sample re-injection.
  • Silanized interior surface prevents adsorption of highly polar molecules.
  • Designed for easy extraction and to maximize content recovery.
  • Attached cap and septum prevent contaminants from entering the sample.
  • Ideal for applications where sample volume is limited or when re-injection of the same sample is needed.
Catalog No. 50-782-510