Learn More
Waters Corp Waters Corporation Autosampler Vial, Deactivated Clear Glass, 12 × 32 mm, 300 μL
Catalog No. 50782536 Shop All Waters Corporation Products
Encompass_Preferred
This Qsert vial is designed for use with limited sample volumes and samples susceptible to adsorption in classic glass vials. It removes the need for a low-volume spring, offering an all-in-one sample collection solution without added parts. Each vial comes with an attached cap and preslit PTFE/silicone septum. They are inspected for cleanliness and held to tight dimensional tolerances.
  • Designed for limited sample volume
  • Suitable for compounds susceptible to adsorption
  • Eliminates the need for a low-volume spring
  • Comes with an attached cap and preslit PTFE/silicone septum
  • Inspected for cleanliness and tight dimensional tolerances
  • All-in-one sample collection solution
  • Includes vial, cap, and septum
  • Ideal for sample preparation applications
Catalog No. 50-782-536 Supplier Waters Corp Supplier No. 186001126DV
This Qsert vial is designed for use with limited sample volumes and samples susceptible to adsorption in classic glass vials. It removes the need for a low-volume spring, offering an all-in-one sample collection solution without added parts. Each vial comes with an attached cap and preslit PTFE/silicone septum. They are inspected for cleanliness and held to tight dimensional tolerances.
  • Designed for limited sample volume
  • Suitable for compounds susceptible to adsorption
  • Eliminates the need for a low-volume spring
  • Comes with an attached cap and preslit PTFE/silicone septum
  • Inspected for cleanliness and tight dimensional tolerances
  • All-in-one sample collection solution
  • Includes vial, cap, and septum
  • Ideal for sample preparation applications