Learn More
Waters Corp WATERS CORPORATION Screw Neck Vial, Deactivated Clear Glass, Qsert, 300 μL
Catalog No. 50782539 Shop All Waters Corporation Products
Encompass_Preferred
Deactivated vials feature a silanized surface that prevents adsorption of highly polar molecules. This treatment with organosilane creates a hydrophobic glass surface, crucial for minimizing sample loss and repeat analysis. The permanent surface alteration ensures these vials can be stored dry indefinitely.
  • Silanized surface prevents adsorption of highly polar molecules.
  • Integrated vial insert (Qsert Vial) eliminates need for a spring and reduces sample preparation time.
  • Permanent surface alteration allows for permanent dry storage.
  • Produced under ISO 9001, ISO 13485, and CGMP standards.
  • Suitable for highly polar substances prone to adsorption.
  • Ideal for biological, pharmaceutical, natural products, insecticides, and herbicides applications.
  • Used for sample preparation.
Catalog No. 50-782-539 Supplier Waters Corp Supplier No. 186001128DV
Deactivated vials feature a silanized surface that prevents adsorption of highly polar molecules. This treatment with organosilane creates a hydrophobic glass surface, crucial for minimizing sample loss and repeat analysis. The permanent surface alteration ensures these vials can be stored dry indefinitely.
  • Silanized surface prevents adsorption of highly polar molecules.
  • Integrated vial insert (Qsert Vial) eliminates need for a spring and reduces sample preparation time.
  • Permanent surface alteration allows for permanent dry storage.
  • Produced under ISO 9001, ISO 13485, and CGMP standards.
  • Suitable for highly polar substances prone to adsorption.
  • Ideal for biological, pharmaceutical, natural products, insecticides, and herbicides applications.
  • Used for sample preparation.