Learn More
Waters Corp Deactivated Clear Glass 12 x 32 mm Snap Neck Max Recovery Vial, 1.5 mL Volume, 100/pk
Catalog No. 50782531 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Deactivated vials have a silanized surface to prevent adsorption of highly polar molecules.
Catalog No. 50-782-531 Supplier Waters Corp Supplier No. 186000984DV
Deactivated vials have a silanized surface to prevent adsorption of highly polar molecules.