Learn More
Air Science Usa Dead Air Box UV 48" Wide Benchtop Enclosure
SDP
Catalog No. 5000580924
Encompass_Preferred
UV Dead Air Boxes / Enclosures Provide a circulation-free workspace designed to protect sensitive samples from contamination. Unlike laminar flow cabinets, these enclosures create a still-air environment with no forced airflow, helping maintain sample integrity. Purair UV Dead Air Boxes are engineered for PCR applications, DNA/RNA work, and other critical laboratory procedures. A built-in 254 nm UV lamp sterilizes the interior and work surface between uses, effectively reducing microorganisms and nucleic acid contamination. Safety interlocks automatically disable UV exposure when the sash is opened. Clear polycarbonate panels provide 360° visibility and allow ambient light into the chamber, while the fully closing front sash minimizes external contamination
Catalog No. 50-005-80924 Supplier Air Science Usa Supplier No. PCR48DBA
UV Dead Air Boxes / Enclosures Provide a circulation-free workspace designed to protect sensitive samples from contamination. Unlike laminar flow cabinets, these enclosures create a still-air environment with no forced airflow, helping maintain sample integrity. Purair UV Dead Air Boxes are engineered for PCR applications, DNA/RNA work, and other critical laboratory procedures. A built-in 254 nm UV lamp sterilizes the interior and work surface between uses, effectively reducing microorganisms and nucleic acid contamination. Safety interlocks automatically disable UV exposure when the sash is opened. Clear polycarbonate panels provide 360° visibility and allow ambient light into the chamber, while the fully closing front sash minimizes external contamination