Learn More
TRIBIOSCIENCE INC Dead Bacterial DNA Eraser, 2 ML
SDP
Catalog No. NC3454598
Encompass_Preferred
Tribioscience's Dead Bacterial DNA Eraser provides an effective and simple method to deplete the dead bacterial DNA and free DNA in the samples. It is enough for 200 sample preparation.
Catalog No. NC3454598 Supplier TRIBIOSCIENCE INC Supplier No. TBS6039
Tribioscience's Dead Bacterial DNA Eraser provides an effective and simple method to deplete the dead bacterial DNA and free DNA in the samples. It is enough for 200 sample preparation.