Learn More
Grainger Dead End Stringer 4 Boxes 60 ft 20A
Catalog No. 502753652
Encompass_Preferred
The KH Industries Dead End Power Stringer is a 15 kV dead end power stringer designed for overhead distribution lines. It features a galvanized steel body with an EPDM rubber housing and stainless steel hardware. The stringer has a 15 kV BIL rating and accommodates conductors from #2 through 556 ACSR. It includes a clevis for attachment to structures and measures 23 inches long by 5 inches wide by 6 inches high. The dead end power stringer provides a method to terminate and dead-end overhead distribution conductors.
Catalog No. 50-275-3652 Supplier Grainger Supplier No. 14A290
The KH Industries Dead End Power Stringer is a 15 kV dead end power stringer designed for overhead distribution lines. It features a galvanized steel body with an EPDM rubber housing and stainless steel hardware. The stringer has a 15 kV BIL rating and accommodates conductors from #2 through 556 ACSR. It includes a clevis for attachment to structures and measures 23 inches long by 5 inches wide by 6 inches high. The dead end power stringer provides a method to terminate and dead-end overhead distribution conductors.