Learn More
Medchemexpress LLC DEAE-dextran | 9015-73-0 | MFCD04334413 | 500,000 Da | 50 G
SDP
Catalog No. 5000283339
Encompass_Preferred
DEAE-dextran (MW 500,000) is a cationic dextran derivative functionalized with diethylaminoethyl groups. Supplied as a powdered reagent for research use, it is commonly used as a transfection agent for nucleic acid delivery and as an ion-exchange material for charge-based separations. CAS 9015-73-0.
  • Cationic polymer suitable for transfection of DNA and RNA.
  • High molecular weight (approx. 500,000 Da) for effective complexation.
  • Soluble in water (approx. 150 mg/mL; may require ultrasonic).
  • Analytical specs include nitrogen content ~3.3% and loss on drying ≤10%.
  • Store sealed at 4°C; solutions stable at -80°C (6 months) or -20°C (1 month).
  • For research use only; not for clinical or human use.
Catalog No. 50-002-83339 Supplier Medchemexpress LLC Supplier No. HYW134327A50G
Add to Cart
Add to Cart
DEAE-dextran (MW 500,000) is a cationic dextran derivative functionalized with diethylaminoethyl groups. Supplied as a powdered reagent for research use, it is commonly used as a transfection agent for nucleic acid delivery and as an ion-exchange material for charge-based separations. CAS 9015-73-0.
  • Cationic polymer suitable for transfection of DNA and RNA.
  • High molecular weight (approx. 500,000 Da) for effective complexation.
  • Soluble in water (approx. 150 mg/mL; may require ultrasonic).
  • Analytical specs include nitrogen content ~3.3% and loss on drying ≤10%.
  • Store sealed at 4°C; solutions stable at -80°C (6 months) or -20°C (1 month).
  • For research use only; not for clinical or human use.