Learn More
Bioworld DEAE SEPAROPOREAGAROSE) 6
Supplier:  Bioworld 201810042
Encompass
DEAE SEPAROPORE (AGAROSE) 6B-CL: BioWorld's DEAE Separopore was prepared by covalent attachment of diethylaminoethyl to Agarose CL-6B. DEAE Separopore is a weak anion exchanger with excellent flow capacities and a high capacity for bindi...
Catalog No. 50-197-664