Learn More
Sigma Aldrich Fine Chemicals Biosciences DEAE SephadexR A 25 Cytiva
Supplier:  Sigma Aldrich Fine Chemicals Biosciences GE17017002
Encompass
DEAE Sephadex(R) A-25 is a weak anion exchanger based on the well documented and well proven Sephadex(R) base matrix.
Catalog No. 50-290-7260