Learn More
Sigma Aldrich Fine Chemicals Biosciences DEAE SephadexR A 25 Cytiva
Catalog No. 502904432
Encompass
DEAE Sephadex(R) A-25 is a weak anion exchanger based on the well documented and well proven Sephadex(R) base matrix.
Catalog No. 50-290-4432 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GE17017001
DEAE Sephadex(R) A-25 is a weak anion exchanger based on the well documented and well proven Sephadex(R) base matrix.