Learn More
Sigma Aldrich Fine Chemicals Biosciences DEAE SepharoseR fast flow
Catalog No. 502900376
Encompass
DEAE Sepharose fast fLow is a weak anion exchanger based on the well established Sepharose(R) fast fLow ion exchange platform extensively used for preparative protein separations in both research and industrial applications.
Catalog No. 50-290-0376 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GE17070910
DEAE Sepharose fast fLow is a weak anion exchanger based on the well established Sepharose(R) fast fLow ion exchange platform extensively used for preparative protein separations in both research and industrial applications.