Learn More
WELCH MATERIALS INC DEAE TanroseTM 6FF, 100mL/bottle
Catalog No. 502093200
Encompass_Preferred
Name: DEAE Tanrose 6FF, specification: 100mL, property: weak anion exchange, bead structure: 6% agarose, mean particle size and range: 90μm, 45-165 μm,ligand density: 110-1600 μmol Cl-/mL, functional group: diethylaminoethyl, flow rate: 50-300 cm/h, pH stability: 2-9(long term), storage condition: 20% ethanol, 4-30°C. Similar Products: DEAE Sepharose FF
Catalog No. 50-209-3200 Supplier WELCH MATERIALS INC Supplier No. Y2034
Name: DEAE Tanrose 6FF, specification: 100mL, property: weak anion exchange, bead structure: 6% agarose, mean particle size and range: 90μm, 45-165 μm,ligand density: 110-1600 μmol Cl-/mL, functional group: diethylaminoethyl, flow rate: 50-300 cm/h, pH stability: 2-9(long term), storage condition: 20% ethanol, 4-30°C. Similar Products: DEAE Sepharose FF