Learn More
Aviva Systems Biology Corp DEAF1 Antibody - N-terminal region (ARP39467_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39467_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DEAF1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human DEAF1. Alternate names: NUDR, SPN, ZMYND5ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rat, Yeast
Catalog No. 50-014-57045