Learn More
Bachem (Glu20)-Glucagon (1-29) (human, rat, porcine) Trifluoroacetate
SDP
Supplier:  Bachem 4079055.1000
Encompass_Preferred
(Glu20)-Glucagon (1-29) (human, rat, porcine) Trifluoroacetate, H-His-Ser-Gln-Gly-Thr-Phe-Thr-Ser-Asp-Tyr-Ser-Lys-Tyr-Leu-Asp-Ser-Arg-Arg-Ala-Glu-Asp-Phe-Val-Gln-Trp-Leu-Met-Asn-Thr-OH, 2022956-46-1, C153H224N42O50S, Mr 3483.78, 1mg. Deamidation product of glucagon.
Catalog No. 50-194-6899