Learn More
Bachem 3-Mercaptopropionyl-Tyr-Phe-Gln-Asn-Cys-Pro-D-Arg-Gly-NH2
SDP
Supplier:  Bachem 4012273.0001
Encompass_Preferred
(Deamino-Cys1,D-Arg8)-Vasopressin Acetate, 3-Mercaptopropionyl-Tyr-Phe-Gln-Asn-Cys-Pro-D-Arg-Gly-NH2, 16679-58-6, C46H64N14O12S2, Mr 1069.23, 1mg. Desmopressin, DDAVP, [Deamino1, DArg8] Vasopressin. Desmopressin (DDAVP), as the first vasopressin analog with a very high and very specific antidiuretic effect, has been widely used for different therapeutic purposes. DDAVP also improves human learning and memory processes. CAS Number (desmopressin acetate): 62288-83-9.
Catalog No. 50-383-373