Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Deapi-platycodin D3 | 67884-05-3 | 1 ML
SDP

Catalog No. 5000417957
Encompass_Preferred

Deapi-platycodin D3 is a triterpenoid saponin derived from the roots of Platycodon grandiflorum. This compound is intended for research use only.

  • Purity: 98.86%
  • Molecular weight: 1255.35
  • Formula: C58H94O29
  • Appearance: White to off-white solid
  • Classification: Terpenoids, triterpenes
  • Initial source: Plants, Campanulaceae, Platycodon grandiflorus (Jacq.) A. DC.
  • Storage (solid): 4°C, protect from light
  • Storage (in solvent): -80°C for 6 months; -20°C for 1 month (protect from light)
  • Solubility (in vitro): DMSO 100 mg/mL (79.66 mM)

Catalog No. 50-004-17957 Supplier Medchemexpress LLC Supplier No. HYN352010MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Deapi-platycodin D3 is a triterpenoid saponin derived from the roots of Platycodon grandiflorum. This compound is intended for research use only.

  • Purity: 98.86%
  • Molecular weight: 1255.35
  • Formula: C58H94O29
  • Appearance: White to off-white solid
  • Classification: Terpenoids, triterpenes
  • Initial source: Plants, Campanulaceae, Platycodon grandiflorus (Jacq.) A. DC.
  • Storage (solid): 4°C, protect from light
  • Storage (in solvent): -80°C for 6 months; -20°C for 1 month (protect from light)
  • Solubility (in vitro): DMSO 100 mg/mL (79.66 mM)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.