Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Deapi-platycodin D3 | 67884-05-3 | 5 MG
SDP

Catalog No. 5000420228
Encompass_Preferred

Deapi-platycodin D3 is a triterpenoid saponin derived from the roots of Platycodon grandiflorum. This product is intended for research use only.

  • Purity of 98.86%
  • Solid appearance
  • White to off-white color
  • Store at 4°C, protected from light; in solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light.
  • Soluble in DMSO at 100 mg/mL (in vitro)

Catalog No. 50-004-20228 Supplier Medchemexpress LLC Supplier No. HYN35205MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Deapi-platycodin D3 is a triterpenoid saponin derived from the roots of Platycodon grandiflorum. This product is intended for research use only.

  • Purity of 98.86%
  • Solid appearance
  • White to off-white color
  • Store at 4°C, protected from light; in solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light.
  • Soluble in DMSO at 100 mg/mL (in vitro)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.