Learn More
Medchemexpress LLC Deapioplatycodin D | 78763-58-3 | 1093.21 | 1 ML
SDP
Catalog No. 5000388910
Encompass_Preferred
Deapioplatycodin D is a triterpenoid saponin isolated from Platycodon grandiflorum, which exhibits anti-HCV (Hepatitis C Virus) activity. It is intended for research use only.
  • HCV inhibitor
  • Isolated from Platycodon grandiflorum
  • Purity: 99.0%
  • Exhibits antiproliferative activity against rat HSC-T6 cells (IC50: 8.24 μM)
Catalog No. 50-003-88910 Supplier Medchemexpress LLC Supplier No. HYN058810MM/1ML
Deapioplatycodin D is a triterpenoid saponin isolated from Platycodon grandiflorum, which exhibits anti-HCV (Hepatitis C Virus) activity. It is intended for research use only.
  • HCV inhibitor
  • Isolated from Platycodon grandiflorum
  • Purity: 99.0%
  • Exhibits antiproliferative activity against rat HSC-T6 cells (IC50: 8.24 μM)