Learn More
Roboz Surgical DEAVER OPERATING SCISSORS
SDP
Catalog No. NC9304668
Encompass
Deaver Scissors; Straight; Sharp-Blunt; 43mm Blade Length; 5-1/2 in. Overall Length
Catalog No. NC9304668 Supplier Roboz Surgical Supplier No. RS6760
Deaver Scissors; Straight; Sharp-Blunt; 43mm Blade Length; 5-1/2 in. Overall Length