Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Deb USA InstantFOAM™ Non-Alcohol Free Hand Sanitizer

Catalog No. 19165454
Encompass_Preferred
Change view
Click to view available options
Volume (Metric):
400 mL
Packaging:
Bottle, one pump
Quantity:
12 Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Packaging Quantity
19-165-454 400 mL Bottle, one pump 12 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-165-454 Supplier Deb USA Supplier No. 55815

missing translation for 'validMainframeMsg'

Instand foam formula sanitizes without drying out skin

  • Unique patented pump and cartridge are ultrasonically sealed and designed to prevent air ingress into the cartridge minimizing the risk of external contamination
  • Foams instantly, spreading product quicker and more effectively
  • Contains skin conditioners to prevent drying of the skin to leave hands feeling soft and supple
  • Perfume-free and dye-free formula will not taint food when used as a hand sanitizer supplemental to handwashing
  • All cartridge components, containers, and packaging are recyclable while formulations are biodegradable
  • Only one dose is required to provide effective hand sanitizing.

Specifications

Product Type Hand Sanitizer
Certifications/Compliance NSF E-3 rated, Federal Food, Drug, and Cosmetic Act
Composition Benzalkonium Chloride, Water, Propylene Glycol, Aloe, Barbadensis Leaf Juice, Cocamidopropyl Betaine, Lauramine Oxide, Tetrasodium EDTA, Magnesium Nitrate, Methylchloroisothiazolinone, Magnesium Chloride, Methylisothiazolinone
For Use With (Application) Ideal for use outside the washroom to provide hand hygiene at the work-point or on the move
Packaging Bottle, one pump
Quantity 12 Pk.
Volume (English) 0.1 gal.
Volume (Metric) 400 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.