Learn More
Deb USA InstantFOAM™ Non-Alcohol Free Hand Sanitizer
Description
- Unique patented pump and cartridge are ultrasonically sealed and designed to prevent air ingress into the cartridge minimizing the risk of external contamination
- Foams instantly, spreading product quicker and more effectively
- Contains skin conditioners to prevent drying of the skin to leave hands feeling soft and supple
- Perfume-free and dye-free formula will not taint food when used as a hand sanitizer supplemental to handwashing
- All cartridge components, containers, and packaging are recyclable while formulations are biodegradable
- Only one dose is required to provide effective hand sanitizing.
Specifications
Specifications
| Product Type | Hand Sanitizer |
| Certifications/Compliance | NSF E-3 rated, Federal Food, Drug, and Cosmetic Act |
| Composition | Benzalkonium Chloride, Water, Propylene Glycol, Aloe, Barbadensis Leaf Juice, Cocamidopropyl Betaine, Lauramine Oxide, Tetrasodium EDTA, Magnesium Nitrate, Methylchloroisothiazolinone, Magnesium Chloride, Methylisothiazolinone |
| For Use With (Application) | Ideal for use outside the washroom to provide hand hygiene at the work-point or on the move |
| Packaging | Bottle, one pump |
| Quantity | 12 Pk. |
| Volume (English) | 0.1 gal. |
| Volume (Metric) | 400 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.