Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Deb USA InstantFOAM™ Non-Alcohol Hand Sanitizer

Catalog No. 19165457
Encompass_Preferred
Change view
Click to view available options
Volume (Metric):
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
19-165-457 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-165-457 Supplier Deb USA Supplier No. 56827

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Alcohol free formula provides effective sanitizing without drying out skin

  • Unique patented pump and cartridge are ultrasonically sealed and designed to prevent air ingress into the cartridge minimizing the risk of external contamination
  • Contains skin conditioners to prevent drying of the skin to leave hands feeling soft and supple
  • All cartridge components, containers, and packaging are recyclable while formulations are biodegradable
  • Only one dose is required to provide effective hand sanitizing

Specifications

Product Type Hand Sanitizer
Color Purple
Form Liquid
Composition Benzalkonium Chloride, Water, Propylene Glycol, Aloe, Barbadensis Leaf Juice, Cocamidopropyl Betaine, Lauramine Oxide, Tetrasodium EDTA, Fragrance, Citric Acid, Magnesium Nitrate, Methylchloroisothiazolinone, Magnesium Chloride, Methylisothiazolinone, blue 1, red 33
For Use With (Application) Kills 99.999% of many common germs in 30 sec.
Packaging Cartridge
pH Range 6.5 to 7.5
Quantity 6 Pk.
Volume (English) 0.26 gal.
Volume (Metric) 1 L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.