Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stoelting™ DeBakey Bulldog Clamp
SDP

Catalog No. 10001392 Shop All Stoelting Co Products
Change view
Click to view available options
Length (English):
3 in.
4.75 in.
Model:
Curved
Straight
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English) Model
10-001-392 4.75 in. Curved
10-001-391 4.75 in. Straight
10-001-385 3 in. Straight
10-001-386 3 in. Curved
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 10-001-392 Supplier Stoelting™ Supplier No. 5212060
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Stoelting™ DeBakey Bulldog Clamp is useful in surgeries that require precise blood stoppage.

One of the most common applications where a bulldog clamp can be used is during coronary surgery where blood flow must be stopped. In general the bulldog clamp, with its many angled varieties and surgical applications, is an essential tool during surgery where blood flow to an organ must be stopped or controlled.

Specifications

For Use With (Application) Neuroscience
Length (English) 4.75 in.
Length (Metric) 12 cm
Model Curved
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.