Learn More
Sklar Instruments TC Debakey Cardiovascular Needle Holder, Sklar, 7 in., Blunt, Straight, Cross-Serrated, Finger Rings / Dual Gold, Tungsten Carbide / Stainless Steel. EA/1.
SDP
Catalog No. 501437770 Shop All Sklar Instruments Products
Encompass_Preferred
TC Debakey Cardiovascular Needle Holder, Sklar, 7 in., Blunt, Straight, Cross-Serrated, Finger Rings / Dual Gold, Tungsten Carbide / Stainless Steel. Sklar's DeBakey Tungsten Carbide Cardiovascular Needle Holder is a ratcheted, finger ring instrument used for holding needles and suturing in cardiovascular procedures. TC instruments are designed to stay sharp for a significantly longer period of time than other standard instruments. Sklar permanently bonds tungsten carbide inserts to the instrument, which maximizes control and precision. TC instruments are identified by gold handles. This product is straight with cross-serrated jaws, blunt/blunt tips and a length of 7 inches.
This item is not returnable. View return policy
Catalog No. 50-143-7770 Supplier Sklar Instruments Supplier No. 218087
TC Debakey Cardiovascular Needle Holder, Sklar, 7 in., Blunt, Straight, Cross-Serrated, Finger Rings / Dual Gold, Tungsten Carbide / Stainless Steel. Sklar's DeBakey Tungsten Carbide Cardiovascular Needle Holder is a ratcheted, finger ring instrument used for holding needles and suturing in cardiovascular procedures. TC instruments are designed to stay sharp for a significantly longer period of time than other standard instruments. Sklar permanently bonds tungsten carbide inserts to the instrument, which maximizes control and precision. TC instruments are identified by gold handles. This product is straight with cross-serrated jaws, blunt/blunt tips and a length of 7 inches.