Learn More
Med Vet International DeBakey Vascular Clamp, 23cm, J0072DV
Catalog No. 5000584272
Encompass_Preferred
Stainless steel. Slightly curved 7.5cm L jaw. DeBakey Atraumatic Vascular Clamp is a ratcheted, finger ring hemostatic clamp used in vascular procedures. The clamp is available in varying sizes by which use is determined by the size of the artery to be held.
Catalog No. 50-005-84272 Supplier Med Vet International Supplier No. JOR72DV
Stainless steel. Slightly curved 7.5cm L jaw. DeBakey Atraumatic Vascular Clamp is a ratcheted, finger ring hemostatic clamp used in vascular procedures. The clamp is available in varying sizes by which use is determined by the size of the artery to be held.