Learn More
TARGETMOL CHEMICALS INC DEBIO-0123 5MG
SDP
Catalog No. 502578947
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Debio-0123 (WEE1-IN-5) is a potent orally available and highly specific WEE1 inhibitor with an IC 50 in the low nanomolar range. Debio-0123 inhibits phospho-CDC2 which translated into an increase in DNA damage and premature entry into mitosis. Debio-0123 increases the antitumoral activity of Carboplatin in vivo whereas neither agent was active alone [1]. purity: 99%
Catalog No. 50-257-8947 Supplier TARGETMOL CHEMICALS INC Supplier No. T98645MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Debio-0123 (WEE1-IN-5) is a potent orally available and highly specific WEE1 inhibitor with an IC 50 in the low nanomolar range. Debio-0123 inhibits phospho-CDC2 which translated into an increase in DNA damage and premature entry into mitosis. Debio-0123 increases the antitumoral activity of Carboplatin in vivo whereas neither agent was active alone [1]. purity: 99%